Learn More
Proteintech ChromoTek TurboGFP-Trap Magnetic Agarose
Product Code. 17323523
Change view
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
17323523 20 Reactions 20 reactions
17303523 10 Reactions 10 reactions
17313523 100 Reactions 100 reactions
17333523 200 Reactions 200 reactions
17343523 400 Reactions 400 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17323523 Supplier Proteintech Supplier No. TBTMA20
TurboGFP-Trap Magnetic Agarose is an affinity reagent for immunoprecipitation (IP) of TurboGFP fusion proteins.
  • Comprises an anti-TurboGFP VHH/ Nanobody coupled to magnetic agarose beads
Specifications
Content And Storage Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year.
For Use With (Application) IP, CoIP, ELISA
Target TurboGFP, CopGFP, Pontellina plumata, maxGFP, maxFP-Green
Label or Dye Magnetic Agarose
Species Alpaca
Sufficient For 20 Reactions