Learn More
Proteintech ChromoTek TurboGFP-Trap Magnetic Agarose
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
Description
- Comprises an anti-TurboGFP VHH/ Nanobody coupled to magnetic agarose beads
Specifications
Specifications
| Content And Storage | Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. |
| For Use With (Application) | IP, CoIP, ELISA |
| Target | TurboGFP, CopGFP, Pontellina plumata, maxGFP, maxFP-Green |
| Label or Dye | Magnetic Agarose |
| Species | Alpaca |
| Sufficient For | 100 Reactions |