Learn More
Proteintech ChromoTek TurboGFP-Trap Agarose Beads
Description
- Comprises an anti-TurboGFP VHH/ Nanobody coupled to agarose beads
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C. Do not freeze. |
| For Use With (Application) | IP, CoIP, ELISA |
| Target | TurboGFP, CopGFP, Pontellina plumata, maxGFP, maxFP-Green |
| Label or Dye | Agarose |
| Species | Alpaca |
| Sufficient For | 10 Reactions |