Learn More
Proteintech ChromoTek Spot-Trap™ Magnetic Particles M-270
Spot-Trap™ Magnetic Particles M-270 are affinity beads for immunoprecipitation (IP) of Spot-tagged proteins.
287.00€ - 7034.00€
Specifications
Content And Storage Upon receipt store at 4°C. Do not freeze.
For Use With (Application) IP, CoIP, ChIP, RIP
Target Spot-Tag Sequence Motif PDRVRAVSHWSS
Label or Dye Magnetic Particles M-270
Species Alpaca
View More Specs
Products 5
Product Code Brand Sufficient For Price Quantity & Availability  
Product Code Brand Sufficient For Price Quantity & Availability  
17313313
View Documents Promotion Details
Proteintech
ETD-10
10 Reactions
287.00€
10 reactions
View alternative products
Save Up to 
»
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
17323313
View Documents Promotion Details
Proteintech
ETD-100
100 Reactions
2151.00€
100 reactions
View alternative products
Save Up to 
»
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
17333313
View Documents Promotion Details
Proteintech
ETD-20
20 Reactions
475.00€
20 reactions
View alternative products
Save Up to 
»
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
17343313
View Documents Promotion Details
Proteintech
ETD-200
200 Reactions
3859.00€
200 reactions
View alternative products
Save Up to 
»
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
17353313
View Documents Promotion Details
Proteintech
ETD-400
400 Reactions
7034.00€
400 reactions
View alternative products
Save Up to 
»
Estimated Shipment: 05-08-2024
Description
  • Comprises an anti-Spot-Tag™ VHH/ Nanobody coupled to Magnetic Particles M-270
Specifications
Specifications
Upon receipt store at 4°C. Do not freeze.
Spot-Tag Sequence Motif PDRVRAVSHWSS
Alpaca
IP, CoIP, ChIP, RIP
Magnetic Particles M-270