missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek Spot-Trap™ Magnetic Particles M-270

Product Code. 17343313
Change view
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
17343313 200 Reactions 200 reactions
17313313 10 Reactions 10 reactions
17323313 100 Reactions 100 reactions
17333313 20 Reactions 20 reactions
17353313 400 Reactions 400 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17343313 Supplier Proteintech Supplier No. etd200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Spot-Trap™ Magnetic Particles M-270 are affinity beads for immunoprecipitation (IP) of Spot-tagged proteins.

  • Comprises an anti-Spot-Tag™ VHH/ Nanobody coupled to Magnetic Particles M-270

Specifications

Content And Storage Upon receipt store at 4°C. Do not freeze.
For Use With (Application) IP, CoIP, ChIP, RIP
Target Spot-Tag Sequence Motif PDRVRAVSHWSS
Label or Dye Magnetic Particles M-270
Species Alpaca
Sufficient For 200 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.