Learn More
Proteintech ChromoTek Spot-Trap™ Agarose
Description
- Comprises an anti-Spot-Tag™ VHH/ Nanobody coupled to agarose beads
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C. Do not freeze. |
| For Use With (Application) | IP, CoIP, ChIP, RIP |
| Target | Spot-Tag Sequence PDRVRAVSHWSS |
| Label or Dye | Agarose |
| Species | Alpaca |
| Sufficient For | 400 Reactions |