Learn More
Proteintech ChromoTek Spot-peptide
Product Code. 17323303
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Product Code. 17323303
Brand: Proteintech EP10
Spot-peptide for elution of Spot-tagged fusion proteins bound to Spot-Cap.
  • Dilute Spot-peptide to 0.1 mM in PBS for elution of Spot-tagged
    proteins from Spot-Cap
  • Shipped at ambient temperature. Upon receipt store at -20°C
  • Aliquot upon reconstitution and store at -20°C
  • Avoid repeated freeze / thaw cycles
  • Reconstitution: Dissolve Spot-peptide in PBS (1 mg Spot-peptide in 140 μL PBS, 10 mg Spot-Peptide in 1.4 mL PBS) to a final concentration of 5 mM (7 mg/mL)
Specifications
Format Lyophilized Powder
Molecular Weight (g/mol) 1397 g/mol
Product Type Peptide of amino acid sequence PDRVRAVSHWSS
Quantity 10 mg