Learn More
Proteintech ChromoTek Spot-Cap™
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
Description
- Spot-Cap is used to purify Spot-tagged proteins from cell extracts of various organisms like mammals, plants, bacteria, yeast, insects etc. Spot-Cap is engineered for gentle elution with Spot-peptide at 4°C and high yields in batch or gravity flow formats.
Specifications
Specifications
| Quantity | 2 mL |
Product Content Correction
Product Title
Spot an opportunity for improvement?Comparta una corrección de contenido