missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek RFP-Trap™ Magnetic Particles M-270
Description
- RFP-Trap Magnetic Particles M-270 is highly recommended, when very large proteins/complexes are investigated, and magnetic separation is needed for IP
- Consists of a RFP VHH/ Nanobody coupled to Magnetic Particles M-270
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C. Do not freeze. |
| For Use With (Application) | IP, CoIP |
| Target | mRFP, mCherry, mRFPruby, mPlum, tagRFP, mKate2, mOrange, PA-mCherry, mScarlet |
| Label or Dye | Magnetic Particles M-270 |
| Species | Alpaca |
| Sufficient For | 100 Reactions |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?