Learn More
Proteintech ChromoTek RFP-Trap™ Magnetic Particles M-270
Description
- RFP-Trap Magnetic Particles M-270 is highly recommended, when very large proteins/complexes are investigated, and magnetic separation is needed for IP
- Consists of a RFP VHH/ Nanobody coupled to Magnetic Particles M-270
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C. Do not freeze. |
| For Use With (Application) | IP, CoIP |
| Target | mRFP, mCherry, mRFPruby, mPlum, tagRFP, mKate2, mOrange, PA-mCherry, mScarlet |
| Label or Dye | Magnetic Particles M-270 |
| Species | Alpaca |
| Sufficient For | 20 Reactions |