Learn More
Proteintech ChromoTek PARP1-Trap Agarose Beads
Product Code. 17373563
Change view
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
17373563 20 Reactions 20 reactions
17353563 10 Reactions 10 reactions
17363563 100 Reactions 100 reactions
17383563 200 Reactions 200 reactions
17393563 400 Reactions 400 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17373563 Supplier Proteintech Supplier No. xta20
PARP1-Trap Agarose is an affinity reagent for IP of human PARP1.
  • Consists of a PARP1 Nanobody/ VHH, which is coupled to agarose beads
Specifications
Content And Storage Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year.
For Use With (Application) IP, CoIP
Target Human PARP1, does not bind to PARP2, 3, and 9
Label or Dye Agarose
Species Alpaca
Sufficient For 20 Reactions