Learn More
Proteintech ChromoTek PARP1-Trap Agarose Beads
Description
- Consists of a PARP1 Nanobody/ VHH, which is coupled to agarose beads
Specifications
Specifications
| Content And Storage | Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. |
| For Use With (Application) | IP, CoIP |
| Target | Human PARP1, does not bind to PARP2, 3, and 9 |
| Label or Dye | Agarose |
| Species | Alpaca |
| Sufficient For | 10 Reactions |