Learn More
Proteintech ChromoTek p53 N-term-Trap Agarose Kit
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Description
- p53-N-term-Trap Agarose is an affinity reagent for IP of p53
- Consists of a p53 Nanobody/ VHH coupled to agarose beads, and which binds to the N-terminus of p53
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |