Learn More
Proteintech ChromoTek p53 N-term-Trap Agarose Kit
Product Code. 17313433
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Product Code. 17313433
Brand: Proteintech ptak20
Kit including 20 reactions of p53 N-term-Trap Agarose and lysis, dilution, wash, and elution buffers.
  • p53-N-term-Trap Agarose is an affinity reagent for IP of p53
  • Consists of a p53 Nanobody/ VHH coupled to agarose beads, and which binds to the N-terminus of p53
Specifications
Quantity 20 Reactions
Sufficient For 20 Reactions