missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek p53 C-term-Trap Agarose

Product Code. 17363423
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
This item is not returnable. View return policy

Product Code. 17363423

Brand: Proteintech pta2100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

p53-C-term-Trap Agarose is an affinity resin for IP of p53.

  • Consists of a p53 Nanobody/ VHH coupled to agarose beads and that binds to the C-terminus of p53

Specifications

Content And Storage Upon receipt store at 4°C. Do not freeze.
Reactivity Tested on human, mouse
For Use With (Application) IP, CoIP
Target p53
Label or Dye Agarose
Species Alpaca
Sufficient For 100 Reactions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.