Learn More
Proteintech ChromoTek p53 C-term-Trap Agarose
Description
- Consists of a p53 Nanobody/ VHH coupled to agarose beads and that binds to the C-terminus of p53
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C. Do not freeze. |
| Reactivity | Tested on human, mouse |
| For Use With (Application) | IP, CoIP |
| Target | p53 |
| Label or Dye | Agarose |
| Species | Alpaca |
| Sufficient For | 10 Reactions |