Learn More
Proteintech ChromoTek Myc-Trap™ Agarose Kit
Description
- Myc-Trap™ Agarose is an affinity reagent for IP and purification of Myc-tagged proteins
- Consists of a Myc Nanobody/ VHH coupled to agarose beads
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |