Learn More
Proteintech ChromoTek mNeonGreen-Trap Agarose Beads
Description
- Consists of a mNeonGreen Nanobody/ VHH coupled to agarose beads
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C. Do not freeze. |
| For Use With (Application) | IP, CoIP, ChIP, RIP |
| Target | mNeonGreen, Branchiostoma lanceolatum |
| Label or Dye | Agarose |
| Species | Alpaca |
| Sufficient For | 20 Reactions |