Learn More
Proteintech ChromoTek MK2-Trap Agarose Kit
Description
- ChromoTek’s MK2-Trap Agarose kit are affinity beads for IP of MK2-fusion proteins
- Comprises a MK2 Nanobody/ VHH conjugated to agarose beads
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |