Learn More
Proteintech ChromoTek MK2-Trap Agarose
Description
- Comprises a MK2 Nanobody/ VHH conjugated to agarose beads
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C. Do not freeze. |
| For Use With (Application) | IP, CoIP |
| Target | K2 (MAP kinase-activated protein kinase 2, MAPKAP-K2); Species-Reactivity: Tested on human, mouse, hamster; Posttranslational modifications: MK2-Trap recognizes unphosphorylated MK2 and Phospho-MK2 (Thr222). Specificity on Phospho-MK2 (Thr334) was not tested. |
| Label or Dye | Agarose |
| Species | Alpaca |
| Sufficient For | 400 Reactions |