missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek MK2-Trap Agarose
Description
- Comprises a MK2 Nanobody/ VHH conjugated to agarose beads
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C. Do not freeze. |
| For Use With (Application) | IP, CoIP |
| Target | K2 (MAP kinase-activated protein kinase 2, MAPKAP-K2); Species-Reactivity: Tested on human, mouse, hamster; Posttranslational modifications: MK2-Trap recognizes unphosphorylated MK2 and Phospho-MK2 (Thr222). Specificity on Phospho-MK2 (Thr334) was not tested. |
| Label or Dye | Agarose |
| Species | Alpaca |
| Sufficient For | 100 Reactions |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?