missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Mdm4/ HdmX-Trap Agarose
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
Description
- Coupled VHH: Recombinant, monoclonal anti-Mdm4/HdmX single domain antibody (sdAb) fragment
Specifications
Specifications
| For Use With (Application) | IP, CoIP |
| Target | Double minute 4 protein (Mdm4/HdmX), human. Note that binding of Mdm4/HdmX-Trap interferes with binding of p53 to Mdm4 HdmX. |
| Label or Dye | Agarose |
| Species | Alpaca |
| Sufficient For | 10 Reactions |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction