Learn More
Proteintech ChromoTek Mdm4/ HdmX-Trap Agarose
Description
- Coupled VHH: Recombinant, monoclonal anti-Mdm4/HdmX single domain antibody (sdAb) fragment
Specifications
Specifications
| For Use With (Application) | IP, CoIP |
| Target | Double minute 4 protein (Mdm4/HdmX), human. Note that binding of Mdm4/HdmX-Trap interferes with binding of p53 to Mdm4 HdmX. |
| Label or Dye | Agarose |
| Species | Alpaca |
| Sufficient For | 20 Reactions |