missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek MBP-Trap Agarose Beads
Description
- Comprise MBP Nanobodies/ VHHs coupled to agarose beads
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C. Do not freeze. |
| For Use With (Application) | IP, CoIP, ChIP, RIP |
| Target | Maltose-binding Protein (MBP) of Escherichia coli |
| Label or Dye | Agarose |
| Species | Alpaca |
| Sufficient For | 10 Reactions |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?