Learn More
Proteintech ChromoTek Histone-Label Atto488
Description
- Consists of an anti-histone VHH conjugated to the fluorescent dye ATTO488. For IF, microscopy of histones, chromosomes, and nuclei
Specifications
Specifications
| Clonality | Monoclonal |
| Quantity | 10 μL |
| For Use With (Application) | Immunofluorescence (IF), Immunocytochemistry (ICC) |
| Conjugate | ATTO488 |
| Target Specificity | Histone H2A-H2B heterodimers |
| Content And Storage | Shipped at ambient temperature. Upon receipt store at +4°C. Stable for 6 month. Do not freeze. Protect from light. |