Description
- Halo-Trap Magnetic Particles M-270 is used to immunoprecipitate Halo-tag proteins from cell extracts of various organisms like mammals, plants, bacteria, yeast, insects etc. in the presence or absence of a covalently bound ligand.
- The interaction between Halo-Trap and the Halo-tag protein is reversible.
Specifications
Specifications
| Binding Capacity | 1.25 μg of recombinant Halo-tag per 25 μL bead slurry |
| Quantity | 20 Reactions |
| For Use With (Application) | IP, CoIP, ChIP, RIP |