Learn More
Proteintech ChromoTek Halo-Trap Agarose Beads
Product Code. 17373403
Change view
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
17373403 200 Reactions 200 reactions
17343403 10 Reactions 10 reactions
17353403 100 Reactions 100 reactions
17363403 20 Reactions 20 reactions
17383403 400 Reactions 400 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17373403 Supplier Proteintech Supplier No. ota200
Halo-Trap Agarose are affinity beads for IP of Halo-tag fusion proteins
  • Comprises a Halo-tag Nanobody /VHH conjugated to agarose beads
Specifications
Content And Storage Upon receipt store at 4°C. Do not freeze.
For Use With (Application) IP, CoIP, ChIP, RIP
Target Halo-tag
Label or Dye Agarose
Species Alpaca
Sufficient For 200 Reactions