Learn More
Proteintech ChromoTek GST-Trap Agarose Kit
Description
- GST-Trap is an affinity resin for IP of GST fusion proteins
- Consists of a GST Nanobody/ VHH coupled to agarose beads
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |