missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek GST-Trap Agarose
Description
- Consists of a GST Nanobody/ VHH coupled to agarose bead
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C. Do not freeze. |
| For Use With (Application) | IP, CoIP, ChIP, RIP |
| Target | Glutathione S-Transferase (GST) from Schistosoma japonicum |
| Label or Dye | Agarose |
| Species | Alpaca |
| Sufficient For | 200 Reactions |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?