Learn More
Proteintech ChromoTek GFP-Trap™ Multiwell Plate
Product Code. 17373363
Change view
Click to view available options
Quantity:
96 Reactions
Unit Size:
96 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17373363 96 Reactions 96 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17373363 Supplier Proteintech Supplier No. gtp96
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
96-well plate coated with GFP VHH, recombinant binding protein.
  • GFP-Trap™ Multiwell Plate for immunoprecipitation (IP) of GFP-tagged proteins
Specifications
Quantity 96 Reactions
Sufficient For 96 Reactions