missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Chromium VI High Range Reagent Water Testing Kit
Click to view available options
Quantity:
100/Pk.
Unit Size:
16g
Specifications
Specifications
| No. of Tests | 100 |
| Quantity | 100/Pk. |
| Range | 0 to 1000mg/L |
| Test Method | Diphenylcarbohydrazide |
| Type | Chromium VI High Range Reagent |
Safety and Handling
Product Identifier
- chromium VI HR reagent
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P404-Store in a closed container.
- MIXTURE LIST-Contains : potassium disulphate, sulphamic acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction