Learn More
Chromium(III) 2-ethylhexanoate, 50% in 2-ethylhexanoic acid
Product Code. 11497877
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497877 250 g 250g
11487877 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11497877 Supplier Thermo Scientific Alfa Aesar Supplier No. A16560.30
Request Bulk or Custom Format
CAS: 3444-17-5 | C24H45CrO6 | 481.61 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3444-17-5
Molecular Formula C24H45CrO6
Molecular Weight (g/mol) 481.61
MDL Number MFCD00014005
InChI Key WBKDDMYJLXVBNI-UHFFFAOYNA-K
Synonym chromium iii 2-ethylhexanoate, chromium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, chromium 3+ salt 3:1, chromic 2-ethylhexanoate, chromium tris 2-ethylhexanoate, chromium 2-ethylhexanoate 1:3, 2-ethylhexanoic acid chromium iii salt, hexanoic acid, 2-ethyl-, chromium 3+ salt, chromium 3+ ; 2-ethylhexanoate
PubChem CID 103012
IUPAC Name chromium(3+);2-ethylhexanoate
SMILES [Cr+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Specifications
Odor Weak, Characteristic
Quantity 250 g
Formula Weight 481.62
Concentration or Composition (by Analyte or Components) 50% in 2-ethylhexanoic acid
Chemical Name or Material Chromium(III) 2-ethylhexanoate