Learn More
JP Selecta™ Chrome Plated Double Burette Clamps
Click to view available options
For Use With (Equipment):
Burettes
Unit Size:
Each
Specifications
Specifications
| Type | Double Burette Clamp |
| Material | Chrome-plated Metal |
| Includes | PVC coated jaws, self adjusting clamp for 2 burettes, with central bosshead screw |
| For Use With (Equipment) | Burettes |
| Length (Metric) | 170 mm |