Learn More
Macherey-Nagel™ Chrombond™ RS 15 C18 ec Polypropylene Flash Column
Click to view available options
:
I.D.: 21.2mm
Specifications
Specifications
| Length (Metric) | 116 mm |
| Material | Polypropylene |
| Type | Flash Column |
| Column Type | Flash Column |
| For Use With (Equipment) | Directly Used in The Teledyne Isco Companion, Rf |