Learn More
Lenz Laborglasinstrumente™ Chromatographic Columns with Frit and NS Socket
Product Code. 10191851
Change view
Click to view available options
Column Capacity:
8 mL
15 mL
23 mL
35 mL
125 mL
430 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Column Capacity Unit Size
10191851 430 mL Each
10121811 125 mL Each
10030511 15 mL Each
10271171 23 mL Each
10668075 8 mL Each
10529581 35 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10191851 Supplier Lenz Laborglasinstrumente™ Supplier No. 05450706
  • ONLINE EXCLUSIVE PRICE
    136.00€
    90.65€ / Pack of 1
    Save 45.35€
    33% Off
View all promotions
Estimated Shipment: 07-12-2026
Specifications
Column Capacity 430 mL
Length (Metric) 600 mm
Material Duran Glass
Diameter (Metric) Outer 30 mm
Includes 0 to 2.5mm Bore PTFE-Hahn Stopcock
Description Chromatography Column
For Use With (Application) Chromatographic applications
Type Chromatography Column
Diameter (Metric) Inner 30 mm