Learn More
Lenz Laborglasintrumente Chromatographic Columns with Frit and Beaded Rim
Click to view available options
Capacity (Metric) Reservoir:
8 mL
35 mL
430 mL
Unit Size:
Each
Description
- Chromatographic columns with sintered frit (porosity: 0)
- With PTFE stopcock
- Made of DURAN™ tubing
Specifications
Specifications
| Capacity (Metric) Reservoir | 430 mL |
| Length (Metric) | 600 mm |
| Material | Duran Glass |
| Diameter (Metric) Outer | 30 mm |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur