missing translation for 'onlineSavingsMsg'
Learn More

Chroman-6-carbaldehyde, 95%, Thermo Scientific™

Product Code. p-3881085
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10113514

Brand: Thermo Scientific Maybridge CC32804CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 55745-97-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD08146589
InChI Key YIHDTNNFJDJYRG-UHFFFAOYSA-N
Synonym chroman-6-carbaldehyde, chromane-6-carbaldehyde, 3,4-dihydro-2h-1-benzopyran-6-carbaldehyde, chroman-6-carboxaldehyde, 3,4-dihydro-2h-chromene-6-carboxaldehyde, 2h-1-benzopyran-6-carboxaldehyde, 3,4-dihydro
PubChem CID 6504215
IUPAC Name 3,4-dihydro-2H-chromene-6-carbaldehyde
SMILES C1CC2=C(C=CC(=C2)C=O)OC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 162.19
Percent Purity 95%
Chemical Name or Material Chroman-6-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.