Learn More
Macherey-Nagel™ Chromafix™ SB Cartridge
Click to view available options
Description:
Adsorbent Weight: 230 mg
Adsorbent Weight: 460 mg
Adsorbent Weight: 920 mg
Unit Size:
Pack of 50
Specifications
Specifications
| Base Material | Silica |
| Description | Adsorbent Weight: 460 mg |
| For Use With (Application) | Organic Acids, Caffeine, Saccharin |
| Particle Size | 45 μm |
| Sorbent | SB |
| Type | Chromafix SB Cartridge |
| Pore Size | 60 Å |
| Quantity | 50 Pack |
| Surface Area (Metric) | 500 m2/g |