Learn More
Macherey-Nagel™ Chromafix™ HR-P Cartridge
Description
Features
- Highly porous polystyrene-divinylbenzene copolymer
- Specific surface 1200 m2/g
- Particle size 50–100 μm
- Very high binding capacity, up to 30 percent of adsorbent weight (for comparison: silica adsorbents about 3 percent)
Recommended application:
- Aromatic compounds
- Phenols from water
- Nitroaromatics from water
- Pesticides from water
- PAHs from oil
Specifications
Specifications
| Bed Weight | 330 mg |
| For Use With (Application) | Aromatic Compounds, Phenols From Water, Nitroaromatics From Water, Pesticides From Water, PAHs From Oil |
| Particle Size | 50 to 100 μm |
| Sorbent | HR-P |
| Type | Chromafix HR-P Cartridge |
| Quantity | 50 Pack |
| Resin Composition | Polystyrene-divinylbenzene adsorbent resin |
| Surface Area (Metric) | 1200 m2/g |