Learn More
Macherey-Nagel™ Chromafix™ Dry (Na2SO4) Cartridge
Product Code. 10639771
Change view
Click to view available options
Bed Weight:
780 mg
1500 mg
2800 mg
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight unitSize
10639771 1500 mg Pack of 50
10639961 2800 mg Pack of 50
10441282 780 mg Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10639771 Supplier Macherey-Nagel™ Supplier No. 731853
In Stock
Specifications
Base Material Dry (Na2 SO4)
Bed Weight 1500 mg
For Use With (Application) Removal of Traces of Water From Organic Solutions
Type Chromafix Dry (Na2 SO4) Cartridge
Quantity 50 Pack