Learn More
Macherey-Nagel™ Chromafix™ Dry (Na2SO4) Cartridge
Click to view available options
Bed Weight:
780 mg
1500 mg
2800 mg
Unit Size:
Pack of 50
Specifications
Specifications
| Base Material | Dry (Na2 SO4) |
| Bed Weight | 780 mg |
| For Use With (Application) | Removal of Traces of Water From Organic Solutions |
| Type | Chromafix Dry (Na2 SO4) Cartridge |
| Quantity | 50 Pack |