Learn More
Thermo Scientific™ Chromacol Vial Racks
Product Code. 11922585
Click to view available options
Holds:
Vials
Vials (100)
Vials (104)
Vials (105)
Material:
Aluminum
Foam
Plastic
For Use With (Equipment):
11 mm OD Vials
12 mm OD Vials
22-23 mm OD Vials
Unit Size:
Each
Pack of 5
Product Code. 11922585
Brand: Thermo Scientific™ T25
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ensure a safe working position on the lab bench and during transport with these robust vial racks.
Save space in the fridge while ensuring a safe working position on the bench and during transport. Available in a various materials, colors and capacities, Thermo Scientific™ Chromacol Vial Racks meet a variety of application needs
Choose from an assortment of chemically resistant and fairly robust racks.
  • Aluminum and plastic models are temperature resistant from -90 to +121°C (-130 to +250°F)
  • Aluminum and plastic models are autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vial Rack
Description Vial Rack
Autoclavable Autoclavable
Color Blue
Holds Vials
Material Aluminum
Positions 25
For Use With (Equipment) 12 mm OD Vials
Limited warranty period:
Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.