Learn More
Macherey-Nagel™ Chromabond™ PTS Separation Column
Click to view available options
Column Capacity (Metric):
1 mL
3 mL
6 mL
15 mL
30 mL
45 mL
70 mL
150 mL
Unit Size:
Pack of 100
Pack of 20
Pack of 50
Description
- Automatic separation of a two-phase mixture without separation funnel
- Two-phase mixtures are completely applied to the column and the phase boundary is determined without further work.
- The special membrane automatically stops the flow when the lower phase has passed.
- The upper phase remains in the column, thus both phases are available for futher analysis.
- Columns must not be run with vacuum or pressure
- PTS for solvents heavier than water, e.g., for trichloromethane, dichloromethane maximum size 150 mL
Specifications
Specifications
| Column Capacity (Metric) | 3 mL |
| Column Type | SPE |
| For Use With (Application) | For Solvents Heavier Than Water, for Trichloromethane, Dichloromethane |
| Sorbent | Chromabond PTS |
| Type | Phase Separation Column |