Learn More
Macherey-Nagel™ Chromabond™ Polypropylene Empty LV SPE Column
Product Code. 10777472
Click to view available options
Bed Weight:
100 mg
200/500 mg
Unit Size:
Pack of 50
Product Code. 10777472
Brand: Macherey-Nagel™ 732501
Only null left
Specifications
Column Capacity (Metric) 15 mL
Column Type Empty LV
Bed Weight 200/500 mg
For Use With (Application) Solid Phase Extraction
Type Empty LV SPE Column
Includes Polyethylene frits