Learn More
Macherey-Nagel™ Chromabond™ PCA Polypropylene SPE Column
Click to view available options
Bed Weight:
500 mg
1 g
Column Capacity (Metric):
3 mL
6 mL
Unit Size:
Pack of 30
Pack of 50
Specifications
Specifications
| Base Material | Silica |
| Column Capacity (Metric) | 3 mL |
| Column Type | SPE |
| Bed Weight | 500 mg |
| For Use With (Application) | Solid Phase Extraction |
| Particle Size | 45 μm |
| Sorbent | Chromabond PCA |
| Type | Solid Phase Extraction Column |
| Pore Size | 60 μm |
| Surface Area (Metric) | 500 m2/g |