Learn More
Macherey-Nagel™ Chromabond™ HR-XC Polypropylene SPE Column
Product Code. 12894431
Click to view available options
Bed Weight:
30 mg
60 mg
150 mg
200 mg
500 mg
Column Capacity (Metric):
1 mL
3 mL
6 mL
Unit Size:
Pack of 30
Product Code. 12894431
Brand: Macherey-Nagel™ 730956
Specifications
Base Material PS/DVB Copolymer
Column Capacity (Metric) 3 mL
Column Type SPE
Bed Weight 60 mg
For Use With (Application) Solid Phase Extraction
Particle Size 85 μm
Sorbent Chromabond HR-XC
Type Solid Phase Extraction Column
Particle Shape Spherical
Pore Size 65 to 75 Å
Surface Area (Metric) 800 m2/g