Learn More
Macherey-Nagel™ Chromabond™ Florisil Polypropylene SPE Column
Specifications
Specifications
| Column Capacity (Metric) | 3 mL |
| Column Type | SPE |
| Bed Weight | 200 mg |
| For Use With (Application) | Solid Phase Extraction |
| Particle Size | 150 to 250 μm |
| Sorbent | Chromabond Florisil |
| Type | Solid Phase Extraction Column |