Learn More
Macherey-Nagel™ Chromabond™ CN/SiOH Polypropylene SPE Column
Product Code. 10584431
Click to view available options
Bed Weight:
500 mg
500 mg/1 g
Column Capacity (Metric):
3 mL
6 mL
Unit Size:
Pack of 30
Pack of 50
Product Code. 10584431
Brand: Macherey-Nagel™ 730112
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Column Capacity (Metric) 3 mL
Column Type SPE
Bed Weight 500 mg
For Use With (Application) Solid Phase Extraction Analysis of PAH
Sorbent Chromabond CN/SiOH
Type Solid Phase Extraction Column
Functionality/Interaction Type p-p Interaction