Learn More
Macherey-Nagel™ Chromabond™ C18 Polypropylene SPE Hydra Column
Click to view available options
Bed Weight:
50 mg
100 mg
200 mg
500 mg
1 g
2 g
3 g
Column Capacity (Metric):
1 mL
3 mL
6 mL
Unit Size:
Pack of 100
Pack of 30
Pack of 50
Specifications
Specifications
| Base Material | Silica |
| Carbon Load | 15% |
| Column Capacity (Metric) | 3 mL |
| Column Type | SPE Hydra |
| Bed Weight | 1 g |
| For Use With (Application) | Solid Phase Extraction of Polar Analytes |
| Particle Size | 45 μm |
| Sorbent | Chromabond C18 |
| Type | SPE Hydra Column |
| Endcapped | No |
| Show More |