Learn More
Macherey-Nagel™ Chromabond™ C18 Fast Flow Polypropylene SPE Column
Click to view available options
Bed Weight:
200 mg
500 mg
Column Capacity (Metric):
3 mL
6 mL
Unit Size:
Pack of 30
Pack of 50
Specifications
Specifications
| Base Material | Silica |
| Carbon Load | 14% |
| Column Capacity (Metric) | 6 mL |
| Column Type | SPE |
| Bed Weight | 500 mg |
| For Use With (Application) | Solid Phase Extraction Non-Polar Compounds |
| Particle Size | 100 μm |
| Sorbent | Chromabond C18 f |
| Type | Solid Phase Extraction Column |
| Endcapped | No |
| Show More |