Learn More
Macherey-Nagel™ Chromabond™ BT 120 SiOH Column
Product Code. 15184202
Change view
Click to view available options
Length (Metric):
255 mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
15184202 255 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15184202 Supplier Macherey-Nagel™ Supplier No. 732965
Only null left
Specifications
Length (Metric) 255 mm
Type Flash Column
Diameter (Metric) Outer 36 mm
For Use With (Equipment) Biotage Isolera Systems